• USA Home
  • 138460 - DL-Arginine hydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
138460 Sigma-Aldrich

DL-Arginine hydrochloride monohydrate

99%

Purchase

Properties

assay   99%
mp   128-130 °C (lit.)
SMILES string   O.Cl.NC(CCCNC(N)=N)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?