• USA Home
  • 139637 - 1-Adamantanamine fumarate

EMAIL THIS PAGE TO A FRIEND
139637 Aldrich

1-Adamantanamine fumarate

96%

Synonym: 1-Aminoadamantane fumarate

  • CAS Number 80789-67-9

  • Empirical Formula (Hill Notation) C14H21NO4

  • Molecular Weight 267.32

  •  MDL number MFCD00074733

Purchase

Properties

assay   96%
mp   264 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?