• USA Home
  • 142263 - 4-Amino-α-diethylamino-o-cresol dihydrochloride

EMAIL THIS PAGE TO A FRIEND
142263 Sigma-Aldrich

4-Amino-α-diethylamino-o-cresol dihydrochloride

97%

Purchase

Properties

InChI Key   PLISWDVQSUTGTR-UHFFFAOYSA-N
assay   97%
mp   215-218 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?