• USA Home
  • 142654 - Ethyl 2-(4-fluorosulfonylphenyl)-4-hydroxy-6-quinolinecarboxylate

EMAIL THIS PAGE TO A FRIEND
142654 Aldrich

Ethyl 2-(4-fluorosulfonylphenyl)-4-hydroxy-6-quinolinecarboxylate

99%

  • CAS Number 80789-71-5

  • Empirical Formula (Hill Notation) C18H14FNO5S

  • Molecular Weight 375.37

  •  MDL number MFCD00006747

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?