• USA Home
  • 143197 - 3-Nitro-1-nitroso-1-propylguanidine

EMAIL THIS PAGE TO A FRIEND
143197 Sigma-Aldrich

3-Nitro-1-nitroso-1-propylguanidine

98%

Synonym: 1-Propyl-3-nitro-1-nitrosoguanidine, N′-Nitro-N-nitroso-N-propylguanidine

Purchase

Properties

assay   98%
mp   108-110 °C (dec.) (lit.)

Description

Features and Benefits

Precursor to 1-diazopropane.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
MF4643000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?