• USA Home
  • 14662 - (+)-1,6-Bis-(2-chlorophenyl)-1,6-diphenyl-2,4-hexadiyne-1,6-diol

EMAIL THIS PAGE TO A FRIEND
14662 Aldrich

(+)-1,6-Bis-(2-chlorophenyl)-1,6-diphenyl-2,4-hexadiyne-1,6-diol

puriss., ≥99.0% (sum of enantiomers, HPLC)

  • CAS Number 86436-19-3

  • Empirical Formula (Hill Notation) C30H20Cl2O2

  • Molecular Weight 483.38

  •  Beilstein Registry Number 8133600

  •  MDL number MFCD00135911

  •  PubChem Substance ID 24848787

Purchase

Properties

grade   puriss.
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +126±2°, c = 1% in methanol
mp   130-133 °C
storage temp.   2-8°C

Description

Other Notes

Resolving agent for compounds lacking functional groups required for the routine resolution by formation of derivatives or salts, e.g. ketones, epoxides, amides, sulfoxides, etc. by formation of clathrates which are recrystallyzed

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?