• USA Home
  • 14670 - (R)-Boc-2-(trifluoromethyl)-β-Phe-OH

EMAIL THIS PAGE TO A FRIEND
14670 Sigma-Aldrich

(R)-Boc-2-(trifluoromethyl)-β-Phe-OH

≥98.0% (HPLC)

Synonym: (R)-3-(Boc-amino)-3-[2-(trifluoromethyl)phenyl]propionic acid, Boc-2-(trifluoromethyl)-L-β-phenylalanine

Purchase

Properties

assay   ≥98.0% (HPLC)
application(s)   peptide synthesis: suitable
SMILES string   CC(C)(C)OC(=O)N[C@H](CC(O)=O)c1ccccc1C(F)(F)F
InChI   1S/C15H18F3NO4/c1-14(2,3)23-13(22)19-11(8-12(20)21)9-6-4-5-7-10(9)15(16,17)18/h4-7,11H,8H2,1-3H3,(H,19,22)(H,20,21)/t11-/m1/s1
InChI key   DYZRXQICSTZZKP-LLVKDONJSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?