EMAIL THIS PAGE TO A FRIEND
146846 Sigma-Aldrich

2-Butoxypyridine

95%

Purchase

Properties

Related Categories Building Blocks, C9, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   OFLSKXBALZCMCX-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.492(lit.)
bp   200 °C(lit.)
density   0.975 g/mL at 25 °C(lit.)

Description

Packaging

25 g in glass bottle

General description

Alkylation of 2-butoxypyridine has been investigated in ionic liquid media.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?