• USA Home
  • 147486 - 6-Chloropiperonyl chloride

EMAIL THIS PAGE TO A FRIEND
147486 Aldrich

6-Chloropiperonyl chloride

96%

Purchase

Properties

assay   96%
mp   64-66 °C (lit.)
Safety & Documentation
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?