• USA Home
  • 147516 - trans-1,2-Cyclohexanedicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
147516 Sigma-Aldrich

trans-1,2-Cyclohexanedicarboxylic acid

95%

Synonym: trans-Cyclohexane-1,2-dicarboxylic acid, trans-Hexahydrophthalic acid

  • CAS Number 2305-32-0

  • Linear Formula C6H10(CO2H)2

  • Molecular Weight 172.18

  •  Beilstein/REAXYS Number 3200609

  •  EC Number 218-975-8

  •  MDL number MFCD00001464

  •  PubChem Substance ID 24848558

  •  NACRES NA.23

Purchase

Properties

Related Categories Carboxylic Acid Monomers, Materials Science, Monomers, Polymer Science
Quality Level   100
assay   95%
mp   228-230 °C (lit.)
SMILES string   OC(=O)[C@@H]1CCCC[C@H]1C(O)=O
InChI   1S/C8H12O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-/m0/s1
InChI key   QSAWQNUELGIYBC-WDSKDSINSA-N

Description

Packaging

25 g in poly bottle

5 g in glass bottle

Other Notes

Remainder cis

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?