• USA Home
  • 14787 - (+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene

EMAIL THIS PAGE TO A FRIEND
14787 Sigma-Aldrich

(+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene

technical, ~90% (GC)

Synonym: (2S,2′S,5S,5′S)-2,2′,5,5′-Tetramethyl-1,1′-(o-phenylene)diphospholane, (S,S)-Me-DUPHOS, (S,S)-Methyl-DUPHOS

  • CAS Number 136735-95-0

  • Empirical Formula (Hill Notation) C18H28P2

  • Molecular Weight 306.36

  •  Beilstein/REAXYS Number 4810602

  •  MDL number MFCD00142322

  •  PubChem Substance ID 24848833

Purchase

Properties

InChI Key   AJNZWRKTWQLAJK-VGWMRTNUSA-N
grade   technical
assay   ~90% (GC)
mp   67-76 °C

Description

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?