• USA Home
  • 14792 - Bis(dimethylamino)methylsilane

EMAIL THIS PAGE TO A FRIEND
14792 Sigma-Aldrich

Bis(dimethylamino)methylsilane

≥95.0% (GC)

  • CAS Number 22705-33-5

  • Empirical Formula (Hill Notation) C5H16N2Si

  • Molecular Weight 132.28

  •  Beilstein/REAXYS Number 2343949

  •  MDL number MFCD00048007

  •  PubChem Substance ID 57647210

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks, Polyamines More...
assay   ≥95.0% (GC)
refractive index   n20/D 1.414
density   0.798 g/mL at 20 °C (lit.)
SMILES string   C[SiH](N(C)C)N(C)C
InChI   1S/C5H16N2Si/c1-6(2)8(5)7(3)4/h8H,1-5H3
InChI key   VBYLGQXERITIBP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?