• USA Home
  • 14847 - Bis(hexamethylene)triamine-pentakis(methylphosphonic acid) solution

EMAIL THIS PAGE TO A FRIEND
14847 Sigma-Aldrich

Bis(hexamethylene)triamine-pentakis(methylphosphonic acid) solution

technical, ~50% in H2O

  • CAS Number 34690-00-1

  • Empirical Formula (Hill Notation) C17H44N3O15P5

  • Molecular Weight 685.41

  •  Beilstein/REAXYS Number 2201286

  •  MDL number MFCD01862934

  •  PubChem Substance ID 24848904

Purchase

Properties

grade   technical
concentration   ~50% in H2O
density   1.24 g/mL at 20 °C
SMILES string   OP(O)(=O)CN(CCCCCCN(CP(O)(O)=O)CP(O)(O)=O)CCCCCCN(CP(O)(O)=O)CP(O)(O)=O
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?