• USA Home
  • 148792 - 4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

EMAIL THIS PAGE TO A FRIEND
148792 Sigma-Aldrich

4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

99%

Synonym: β,4-Dihydroxy-3-methoxyphenethyl alcohol piperidinium salt, 3-Methoxy-4-hydroxy-phenylglycol piperazine, 4-Hydroxy-3-methoxyphenylglycol hemipiperazine salt, 4-Hydroxy-3-methoxyphenylglycol piperazine;, MHPG piperazine;, MOPEG piperazine;, MOPEG piperidinium salt, MOPEG

Purchase

Properties

assay   99%
mp   116-120 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?