• USA Home
  • 149802 - 6-Hydroxydopamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
149802 Sigma-Aldrich

6-Hydroxydopamine hydrochloride

98%

Synonym: 2,4,5-Trihydroxyphenethylamine hydrochloride, 2,5-Dihydroxytyramine hydrochloride, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrochloride, 6-OHDA

  • CAS Number 28094-15-7

  • Linear Formula (HO)3C6H2CH2CH2NH2 · HCl

  • Molecular Weight 205.64

  •  Beilstein/REAXYS Number 4274007

  •  EC Number 248-837-2

  •  MDL number MFCD00012895

Purchase

Properties

assay   98%
mp   232-233 °C (dec.) (lit.)
SMILES string   Cl.NCCc1cc(O)c(O)cc1O
InChI   1S/C8H11NO3.ClH/c9-2-1-5-3-7(11)8(12)4-6(5)10;/h3-4,10-12H,1-2,9H2;1H
InChI key   QLMRJHFAGVFUAC-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Neurotoxin that destroys catecholaminergic terminals.

Reconstitution

Solutions should be freshly prepared and protected from exposure to light. Solutions turn red as it oxidizes.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DC4650000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H4381 6-Hydroxydopamine hydrochloride, ≥97% (titration), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?