EMAIL THIS PAGE TO A FRIEND
14985 Sigma-Aldrich

Boc-D-Phe(3-CN)-OH

≥98.0% (HPLC)

Synonym: Boc-3-cyano-D-phenylalanine

  • CAS Number 205445-56-3

  • Empirical Formula (Hill Notation) C15H18N2O4

  • Molecular Weight 290.31

  •  MDL number MFCD00797561

  •  PubChem Substance ID 57647244

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0% (HPLC)
SMILES string   CC(C)(C)OC(=O)N[C@H](Cc1cccc(c1)C#N)C(O)=O
InChI   1S/C15H18N2O4/c1-15(2,3)21-14(20)17-12(13(18)19)8-10-5-4-6-11(7-10)9-16/h4-7,12H,8H2,1-3H3,(H,17,20)(H,18,19)/t12-/m1/s1
InChI key   FDQDHMZKOPOWFE-GFCCVEGCSA-N

Description

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?