EMAIL THIS PAGE TO A FRIEND
14995 Sigma-Aldrich

Boc-D-Phe(3-F)-OH

≥98.0% (TLC)

Synonym: Boc-3-fluoro-D-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry | Peptide Synthesis, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   FPCCREICRYPTTL-LLVKDONJSA-N
assay   ≥98.0% (TLC)
optical activity   [α]20/D −6.5±1°, c = 1% in methanol

Description

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?