EMAIL THIS PAGE TO A FRIEND
14997 Sigma-Aldrich

Boc-D-Phe(2-Me)-OH

≥98.0% (TLC)

Synonym: Boc-2-methyl-D-phenylalanine

  • CAS Number 80102-29-0

  • Empirical Formula (Hill Notation) C15H21NO4

  • Molecular Weight 279.33

  •  MDL number MFCD00671403

  •  PubChem Substance ID 57647254

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0% (TLC)
application(s)   peptide synthesis: suitable
storage temp.   room temp
SMILES string   Cc1ccccc1C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
InChI   1S/C15H21NO4/c1-10-7-5-6-8-11(10)9-12(13(17)18)16-14(19)20-15(2,3)4/h5-8,12H,9H2,1-4H3,(H,16,19)(H,17,18)/t12-/m1/s1
InChI key   PCGOCPOJLMLJAR-GFCCVEGCSA-N

Description

Packaging

1 g in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?