EMAIL THIS PAGE TO A FRIEND
15181 Sigma-Aldrich

Boc-D-Phe-ONp

≥98.0%

Synonym: Boc-D-phenylalanine 4-nitrophenyl ester

  • CAS Number 16159-70-9

  • Empirical Formula (Hill Notation) C20H22N2O6

  • Molecular Weight 386.40

  •  Beilstein/REAXYS Number 5773035

  •  MDL number MFCD00069874

  •  PubChem Substance ID 24848857

Purchase

Properties

assay   ≥98.0%
storage temp.   2-8°C
SMILES string   CC(C)(C)OC(=O)N[C@H](Cc1ccccc1)C(=O)Oc2ccc(cc2)[N+]([O-])=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?