EMAIL THIS PAGE TO A FRIEND
15182 Sigma-Aldrich

Boc-D-Ser-OH

≥98.0% (TLC)

Synonym: Boc-D-serine

  • CAS Number 6368-20-3

  • Empirical Formula (Hill Notation) C8H15NO5

  • Molecular Weight 205.21

  •  Beilstein/REAXYS Number 1874714

  •  MDL number MFCD00063142

  •  PubChem Substance ID 57647334

  •  NACRES NA.22

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Serine - Amino Acids and Derivatives More...
assay   ≥98.0% (TLC)
SMILES string   CC(C)(C)OC(=O)N[C@H](CO)C(O)=O
InChI   1S/C8H15NO5/c1-8(2,3)14-7(13)9-5(4-10)6(11)12/h5,10H,4H2,1-3H3,(H,9,13)(H,11,12)/t5-/m1/s1
InChI key   FHOAKXBXYSJBGX-RXMQYKEDSA-N

Description

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?