EMAIL THIS PAGE TO A FRIEND
15187 Sigma-Aldrich

Boc-D-Tyr-OH

≥98.0% (HPLC)

Synonym: Boc-D-tyrosine

  • CAS Number 70642-86-3

  • Empirical Formula (Hill Notation) C14H19NO5

  • Molecular Weight 281.30

  •  Beilstein/REAXYS Number 3085467

  •  MDL number MFCD00063030

  •  eCl@ss 32160406

  •  PubChem Substance ID 57647337

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Tyrosine More...
InChI Key   CNBUSIJNWNXLQQ-LLVKDONJSA-N
assay   ≥98.0% (HPLC)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?