EMAIL THIS PAGE TO A FRIEND
152439 Sigma-Aldrich

Dibutyl phthalate

ReagentPlus®, ≥99%

Synonym: n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

  • CAS Number 84-74-2

  • Linear Formula C6H4-1,2-[CO2(CH2)3CH3]2

  • Molecular Weight 278.34

  •  Beilstein/REAXYS Number 1914064

  •  EC Number 201-557-4

  •  MDL number MFCD00009441

  •  PubChem Substance ID 24849229

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
Quality Level   100
grade   ReagentPlus®
vapor density   9.6 (vs air)
vapor pressure   1 mmHg ( 147 °C)
assay   ≥99%
form   liquid
autoignition temp.   756 °F
expl. lim.   0.47 %, 236 °F
color   APHA: ≤10
refractive index   n20/D 1.492 (lit.)
bp   340 °C (lit.)
mp   −35 °C (lit.)
density   1.043 g/mL at 25 °C (lit.)
SMILES string   CCCCOC(=O)c1ccccc1C(=O)OCCCC
InChI   1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3
InChI key   DOIRQSBPFJWKBE-UHFFFAOYSA-N
Gene Information   mouse ... Esr1(13982)

Description

Application

Plasticizer

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
TI0875000
Flash Point(F) 
339.8 °F
Flash Point(C) 
171 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?