• USA Home
  • 153206 - (±)-3-Benzylphthalide

EMAIL THIS PAGE TO A FRIEND
153206 Aldrich

(±)-3-Benzylphthalide

98%

  • CAS Number 7011-98-5

  • Empirical Formula (Hill Notation) C15H12O2

  • Molecular Weight 224.25

  •  MDL number MFCD00005914

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?