• USA Home
  • 153478 - 2,3,4-Trichlorophenol

EMAIL THIS PAGE TO A FRIEND
153478 Sigma-Aldrich

2,3,4-Trichlorophenol

99%

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
assay   99%
mp   75-79 °C (lit.)
SMILES string   Oc1ccc(Cl)c(Cl)c1Cl
InChI   1S/C6H3Cl3O/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H
InChI key   HSQFVBWFPBKHEB-UHFFFAOYSA-N

Description

General description

2,3,4-Trichlorophenol was determined in human urine by ultra-high-performance liquid chromatography-tandem mass spectrometry (UPLC-MS-MS).

Application

2,3,4-Trichlorophenol was used to study the catalytic activity of mimetic enzymes instead of peroxidase.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?