EMAIL THIS PAGE TO A FRIEND
15351 Sigma-Aldrich

Boc-D-Phe(4-F)-OH

≥99.0% (TLC)

Synonym: Boc-4-fluoro-D-phenylalanine

  • CAS Number 57292-45-2

  • Empirical Formula (Hill Notation) C14H18FNO4

  • Molecular Weight 283.30

  •  Beilstein/REAXYS Number 4323475

  •  MDL number MFCD00079669

  •  PubChem Substance ID 57647389

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
assay   ≥99.0% (TLC)
application(s)   peptide synthesis: suitable
mp   83-86 °C
SMILES string   CC(C)(C)OC(=O)N[C@H](Cc1ccc(F)cc1)C(O)=O
InChI   1S/C14H18FNO4/c1-14(2,3)20-13(19)16-11(12(17)18)8-9-4-6-10(15)7-5-9/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)/t11-/m1/s1
InChI key   RCXSXRAUMLKRRL-LLVKDONJSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?