• USA Home
  • 15427 - Boc-His(Boc)-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
15427 Sigma-Aldrich

Boc-His(Boc)-OH (dicyclohexylammonium) salt

≥99.0% (NT)

Synonym: Nα,N(im)-di-Boc-L-histidine (dicyclohexylammonium) salt

  • CAS Number 31687-58-8

  • Empirical Formula (Hill Notation) C16H25N3O6 · C12H23N

  • Molecular Weight 536.70

  •  Beilstein/REAXYS Number 4120943

  •  EC Number 250-764-6

  •  MDL number MFCD06804675

  •  PubChem Substance ID 24849420

Purchase

Properties

assay   ≥99.0% (NT)
optical activity   [α]20/D +19±1°, c = 2% in chloroform
mp   160-165 °C (dec.)
SMILES string   C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N[C@@H](Cc3cn(cn3)C(=O)OC(C)(C)C)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?