EMAIL THIS PAGE TO A FRIEND
15437 Sigma-Aldrich

Boc-Glu(OcHx)-OH

purum, ≥98.0% (sum of enantiomers, TLC)

Synonym: Boc-L-glutamic acid γ-cyclohexyl ester, Boc-L-glutamic acid 5-cyclohexyl ester

  • CAS Number 73821-97-3

  • Linear Formula C6H11OCOCH2CH2CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 329.39

  •  Beilstein/REAXYS Number 3595818

  •  MDL number MFCD00065570

  •  PubChem Substance ID 24849433

Purchase

Properties

grade   purum
assay   ≥98.0% (sum of enantiomers, TLC)
optical activity   [α]20/D −16.5±1°, c = 2% in DMF
impurities   ≤5% solvent
mp   54-57 °C
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Natural Amino Acid Building Blocks for Peptide Synthesis

With a growing peptide drug market the fast, reliable and uncomplicated synthesis of peptides is of paramount importance.1 Today, the most common synthetic approaches to medium and even large peptide...
Matthias Junkers
ChemFiles 2008, 8.7, 22.
Keywords: Building blocks, Peptide synthesis, Peptidomimetics, Solid phase peptide synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?