EMAIL THIS PAGE TO A FRIEND
15536 Sigma-Aldrich

Boc-Lys(2-Br-Z)-OH

≥98.0% (TLC)

Synonym: Nα-Boc-Nε-(2-bromo-Z)-L-lysine

  • CAS Number 47592-74-5

  • Empirical Formula (Hill Notation) C19H27BrN2O6

  • Molecular Weight 459.33

  •  MDL number MFCD00076962

  •  PubChem Substance ID 57647481

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Synthesis and Peptide Chemistry More...
Quality Level   100
assay   ≥98.0% (TLC)
application(s)   peptide synthesis: suitable
mp   72-76 °C
SMILES string   CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCc1ccccc1Br)C(O)=O
InChI   1S/C19H27BrN2O6/c1-19(2,3)28-18(26)22-15(16(23)24)10-6-7-11-21-17(25)27-12-13-8-4-5-9-14(13)20/h4-5,8-9,15H,6-7,10-12H2,1-3H3,(H,21,25)(H,22,26)(H,23,24)/t15-/m0/s1
InChI key   IBYVEZVZHJKJOU-HNNXBMFYSA-N

Description

Packaging

1 g in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?