• USA Home
  • 155594 - 2,4,6-Trimethoxybenzonitrile

EMAIL THIS PAGE TO A FRIEND
155594 Sigma-Aldrich

2,4,6-Trimethoxybenzonitrile

98%

Purchase

Properties

Related Categories Building Blocks, C10 to C27, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
assay   98%
mp   143-145 °C (lit.)
SMILES string   COc1cc(OC)c(C#N)c(OC)c1
InChI   1S/C10H11NO3/c1-12-7-4-9(13-2)8(6-11)10(5-7)14-3/h4-5H,1-3H3
InChI key   GBRHJUMDNWLSCT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?