• USA Home
  • 155616 - (S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

EMAIL THIS PAGE TO A FRIEND
155616 Sigma-Aldrich

(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

98%

Synonym: (−)-MTPA, Mosher’s acid

  • CAS Number 17257-71-5

  • Linear Formula C6H5C(OCH3)(CF3)CO2H

  • Molecular Weight 234.17

  •  Beilstein Registry Number 4684048

  •  EC Number 241-292-1

  •  MDL number MFCD00064200

Purchase

Properties

InChI Key   JJYKJUXBWFATTE-VIFPVBQESA-N
assay   98%
optical activity   [α]20/D −70°, c = 1.6 in methanol
optical purity   ee: 95% (GLC)
refractive index   n20/D 1.474(lit.)
bp   95-97 °C/0.05 mmHg(lit.)
mp   46-49 °C (lit.)
density   1.303 g/mL at 25 °C(lit.)

Description

Application

Derivatizing reagent for determination of enantiomeric excess by NMR.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

318035 (S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?