• USA Home
  • 157899 - 2,4-Dimethyl-3-pentanol

EMAIL THIS PAGE TO A FRIEND
157899 Sigma-Aldrich

2,4-Dimethyl-3-pentanol

technical grade, 94%

Synonym: Diisopropylcarbinol

  • CAS Number 600-36-2

  • Linear Formula (CH3)2CHCH(OH)CH(CH3)2

  • Molecular Weight 116.20

  •  Beilstein/REAXYS Number 1731593

  •  MDL number MFCD00008921

Purchase

Properties

grade   technical grade
InChI Key   BAYAKMPRFGNNFW-UHFFFAOYSA-N
assay   94%
refractive index   n20/D 1.425 (lit.)
bp   139-140 °C(lit.)
density   0.829 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?