EMAIL THIS PAGE TO A FRIEND
158526 Sigma-Aldrich

Ammonium acetate

98%

Purchase

Properties

assay   98%
form   solid
mp   110-112 °C (dec.) (lit.)
SMILES string   N.CC(O)=O
InChI   1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key   USFZMSVCRYTOJT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
AF3675000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A7262 Ammonium acetate, reagent grade, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?