EMAIL THIS PAGE TO A FRIEND
160539 Sigma-Aldrich

Ethidium bromide

95%

Synonym: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, EtBr, Homidium bromide

  • CAS Number 1239-45-8

  • Empirical Formula (Hill Notation) C21H20BrN3

  • Molecular Weight 394.31

  •  Beilstein/REAXYS Number 3642536

  •  EC Number 214-984-6

  •  MDL number MFCD00011724

Purchase

Properties

assay   95%
mp   260-262 °C (dec.) (lit.)
λmax   506 nm
SMILES string   [Br-].CC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14
InChI   1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H
InChI key   ZMMJGEGLRURXTF-UHFFFAOYSA-N

Description

Application

Ethidium bromide (EtBr) is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the background is not necessary with a low stain concentration (10 μg/ml). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids. It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA. Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase.

Frameshift mutagen which intercalates double-stranded DNA and RNA.

Frameshift mutagen which intercalates double-stranded DNA and RNA.

Biochem/physiol Actions

Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

Reconstitution

For staining a gel after electrophoresis, dilute a sample of the stock solution to 0.5 μg/ml with water and incubate the gel for 15-30 min. Destaining is usually not needed but can be carried out in water for 15 min if decreased background is necessary. The DNA bands can then be detected on a UV light box (254 nm wavelength). Ethidium bromide can also be incorporated into the gel and running buffer at 0.5 μg/ml and visualized immediately after electrophoresis.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811PIH 6.1 / PGI
WGK Germany 
3
RTECS 
SF7950000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

01494 Nancy-520, suitable for RT-PCR, ≥97.0% (HPCE)

replaced by

Product #

Description

Add to Cart

E8751 Ethidium bromide, ~95% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?