EMAIL THIS PAGE TO A FRIEND
160881 Sigma-Aldrich

Resolve-Al Pr

99%

Synonym: Pr(dpm)3, Pr(thd)3, Pr(tmhd)3, Praseodymium tris(dipivaloylmethane), Praseodymium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate), Praseodymium-shift, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium

Purchase

Properties

assay   99%
mp   219-221 °C (lit.)
SMILES string   CC(C)(C)C(=O)\C=C(/O[Pr](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

Description

Legal Information

Resolve-Al is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?