• USA Home
  • 161861 - 2-(2,4,5-Trichlorophenoxy)propionic acid

EMAIL THIS PAGE TO A FRIEND
161861

2-(2,4,5-Trichlorophenoxy)propionic acid

97%

Synonym: 2,4,5-TP, Fenoprop, Silvex®

  • CAS Number 93-72-1

  • Empirical Formula (Hill Notation) C9H7Cl3O3

  • Molecular Weight 269.51

  •  Beilstein/REAXYS Number 1985768

  •  MDL number MFCD00002646

Purchase

Properties

assay   97%
SMILES string   CC(Oc1cc(Cl)c(Cl)cc1Cl)C(O)=O
InChI   1S/C9H7Cl3O3/c1-4(9(13)14)15-8-3-6(11)5(10)2-7(8)12/h2-4H,1H3,(H,13,14)
InChI key   ZLSWBLPERHFHIS-UHFFFAOYSA-N

Description

Legal Information

Silvex is a registered trademark of Silberline Manufacturing Co., Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?