• USA Home
  • 161926 - 5,5-Diphenylhydantoin

EMAIL THIS PAGE TO A FRIEND
161926 Sigma-Aldrich

5,5-Diphenylhydantoin

≥99%

Synonym: 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

  • CAS Number 57-41-0

  • Empirical Formula (Hill Notation) C15H12N2O2

  • Molecular Weight 252.27

  •  Beilstein/REAXYS Number 384532

  •  EC Number 200-328-6

  •  MDL number MFCD00005264

Purchase

Properties

assay   ≥99%
mp   293-295 °C (lit.)
SMILES string   O=C1NC(=O)C(N1)(c2ccccc2)c3ccccc3
InChI   1S/C15H12N2O2/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H2,16,17,18,19)
InChI key   CXOFVDLJLONNDW-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
MU1050000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D4007 5,5-Diphenylhydantoin, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?