• USA Home
  • 162264 - Tris(2-methylallyl)amine

EMAIL THIS PAGE TO A FRIEND
162264 Sigma-Aldrich

Tris(2-methylallyl)amine

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.4575 (lit.)
bp   83-85 °C/15 mmHg (lit.)
density   0.794 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?