EMAIL THIS PAGE TO A FRIEND
162612 Aldrich

D-Phenylalanine

≥99%

Synonym: (R)-(+)-Phenylalanine, (R)-2-Amino-3-phenylpropionic acid, D-2-Amino-3-phenylpropanoic acid

  • CAS Number 673-06-3

  • Linear Formula C6H5CH2CH(NH2)CO2H

  • Molecular Weight 165.19

  •  Beilstein Registry Number 2804068

  •  EC Number 211-603-5

  •  MDL number MFCD00004270

  •  PubChem Substance ID 329751170

Purchase

Properties

InChI Key   COLNVLDHVKWLRT-MRVPVSSYSA-N
assay   ≥99%
optical activity   [α]23/D +35°, c = 2 in H2O
optical purity   ee: 98% (GLC)
mp   273-276 °C(lit.)
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
AY7533000
Protocols & Articles

Articles

HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P1751 D-Phenylalanine, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?