EMAIL THIS PAGE TO A FRIEND
162728 Sigma-Aldrich

DL-Leucine

≥99%

Synonym: (±)-Amino-4-methylpentanoic acid

  • CAS Number 328-39-2

  • Linear Formula (CH3)2CHCH2CH(NH2)CO2H

  • Molecular Weight 131.17

  •  Beilstein/REAXYS Number 636005

  •  EC Number 206-328-2

  •  MDL number MFCD00063087

Purchase

Properties

assay   ≥99%
mp   293-296 °C (subl.) (lit.)
SMILES string   CC(C)CC(N)C(O)=O
InChI   1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)
InChI key   ROHFNLRQFUQHCH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

L7875 DL-Leucine, ≥99% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?