• USA Home
  • 164453 - 2-Thiophenecarboxamide

EMAIL THIS PAGE TO A FRIEND
164453 Sigma-Aldrich

2-Thiophenecarboxamide

99%

  • CAS Number 5813-89-8

  • Empirical Formula (Hill Notation) C5H5NOS

  • Molecular Weight 127.16

  •  MDL number MFCD00010425

  •  PubChem Substance ID 24850074

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   DENPQNAWGQXKCU-UHFFFAOYSA-N
assay   99%
mp   181-183 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XM8290000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?