• USA Home
  • 16478 - syn-7-Bromobicyclo[2.2.1]hept-2-ene

EMAIL THIS PAGE TO A FRIEND
16478 Sigma-Aldrich

syn-7-Bromobicyclo[2.2.1]hept-2-ene

≥97.0% (GC)

  • CAS Number 20047-65-8

  • Empirical Formula (Hill Notation) C7H9Br

  • Molecular Weight 173.05

  •  Beilstein/REAXYS Number 2039758

  •  MDL number MFCD01862962

  •  PubChem Substance ID 57647566

Purchase

Properties

Related Categories Alkenyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
assay   ≥97.0% (GC)
density   1.46 g/mL at 20 °C (lit.)
storage temp.   room temp
SMILES string   Br[C@@H]1[C@H]2CC[C@@H]1C=C2
InChI   1S/C7H9Br/c8-7-5-1-2-6(7)4-3-5/h1-2,5-7H,3-4H2/t5-,6+,7-
InChI key   XFPAUZYYSFLZRD-KVSKUHBBSA-N

Description

Application

syn-7-Bromobicyclo[2.2.1]hept-2-ene was used in the synthesis of syn- and anti-7-(1,2-butadienyl)bicyclo[2.2.1]hept-2-ene.

Packaging

5 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?