• USA Home
  • 167371 - 3-tert-Butyl-4-hydroxyanisole

EMAIL THIS PAGE TO A FRIEND
167371 Aldrich

3-tert-Butyl-4-hydroxyanisole

≥95%

Synonym: 2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA

  • CAS Number 121-00-6

  • Empirical Formula (Hill Notation) C11H16O2

  • Molecular Weight 180.24

  •  Beilstein Registry Number 1867499

  •  EC Number 204-442-7

  •  MDL number MFCD00040484

Purchase

Properties

InChI Key   MRBKEAMVRSLQPH-UHFFFAOYSA-N
assay   ≥95%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SK1575000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B1253 Butylated hydroxyanisole, ≥98.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?