• USA Home
  • 16749 - (S)-6-Chloromethyl-5,6-dihydro-pyran-2-one

EMAIL THIS PAGE TO A FRIEND
16749 Sigma-Aldrich

(S)-6-Chloromethyl-5,6-dihydro-pyran-2-one

≥98.0%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Pyrans More...
assay   ≥98.0%
optical activity   [α]/D −162.0±2.0, c = 0.5 in chloroform
SMILES string   ClC[C@@H]1CC=CC(=O)O1
InChI   1S/C6H7ClO2/c7-4-5-2-1-3-6(8)9-5/h1,3,5H,2,4H2/t5-/m0/s1
InChI key   NVMBKTXDBXNDQK-YFKPBYRVSA-N

Description

Packaging

1 g in poly tube

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?