EMAIL THIS PAGE TO A FRIEND
167649 Sigma-Aldrich

Hydrobenzamide

98%

Synonym: N,N′-Dibenzylidenetoluene-α,α-diamine

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
assay   98%
mp   102-105 °C (lit.)
storage temp.   room temp
SMILES string   c1ccc(cc1)\C=N\C(\N=C\c2ccccc2)c3ccccc3
InChI   1S/C21H18N2/c1-4-10-18(11-5-1)16-22-21(20-14-8-3-9-15-20)23-17-19-12-6-2-7-13-19/h1-17,21H/b22-16+,23-17+
InChI key   VUYRFIIFTJICNA-LKNRODPVSA-N

Description

General description

Hydrobenzamide is the key intermediate in the three-component reaction of aldehydes, alkyl bromides and ammonia. It was the intermediate during the synthesis of racemic1,2-diphenyl-1,2-diaminoethane.

Application

Hydrobenzamide was used in the synthesis of symmetrical iminobisphosphonic acids.

Packaging

50 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?