• USA Home
  • 168491 - 2-(Dibutylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
168491 Aldrich

2-(Dibutylamino)ethanol

99%

Synonym: N,N-Dibutylethanolamine

  • CAS Number 102-81-8

  • Linear Formula (CH3CH2CH2CH2)2NCH2CH2OH

  • Molecular Weight 173.30

  •  Beilstein Registry Number 1738441

  •  EC Number 203-057-1

  •  MDL number MFCD00014033

Purchase

Properties

vapor density   6 (vs air)
InChI Key   IWSZDQRGNFLMJS-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.444(lit.)
bp   229-230 °C(lit.)
density   0.86 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2873 6.1 / PGIII
WGK Germany 
1
RTECS 
KK3850000
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

550035 2-(Dibutylamino)ethanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?