EMAIL THIS PAGE TO A FRIEND
171255 Sigma-Aldrich

Oxalacetic acid

98%

Synonym: 2-Oxosuccinic acid, Oxobutanedioic acid

  • CAS Number 328-42-7

  • Linear Formula HO2CCH2COCO2H

  • Molecular Weight 132.07

  •  Beilstein/REAXYS Number 1705475

  •  EC Number 206-329-8

  •  MDL number MFCD00002592

Purchase

Properties

assay   98%
mp   161 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O4126 Oxaloacetic acid, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?