• USA Home
  • 172723 - α,α,α-Tribromoquinaldine

EMAIL THIS PAGE TO A FRIEND
172723 Sigma-Aldrich

α,α,α-Tribromoquinaldine

≥98%

Purchase

Properties

assay   ≥98%
mp   127-129 °C (lit.)
SMILES string   BrC(Br)(Br)c1ccc2ccccc2n1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VC3250750

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?