EMAIL THIS PAGE TO A FRIEND
173444 Sigma-Aldrich

DL-2-Allylglycine

99%

Synonym: (±)-2-Amino-4-pentenoic acid, DL-C-Allylglycine

  • CAS Number 7685-44-1

  • Linear Formula H2C=CHCH2CH(NH2)CO2H

  • Molecular Weight 115.13

  •  Beilstein/REAXYS Number 1748645

  •  EC Number 231-689-8

  •  MDL number MFCD00063103

Purchase

Properties

InChI Key   WNNNWFKQCKFSDK-UHFFFAOYSA-N
assay   99%
mp   258-260 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A8378 DL-2-Allylglycine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?