• USA Home
  • 17376 - 4-Hydroxy-2,5,6-triaminopyrimidine sulfate salt

EMAIL THIS PAGE TO A FRIEND
17376 Sigma-Aldrich

4-Hydroxy-2,5,6-triaminopyrimidine sulfate salt

technical, ≥97.0% (T)

Synonym: 2,5,6-Triamino-4-pyrimidol sulfate salt

  • CAS Number 35011-47-3

  • Empirical Formula (Hill Notation) C4H7N5O · H2SO4

  • Molecular Weight 239.21

  •  Beilstein/REAXYS Number 4621048

  •  MDL number MFCD00012785

  •  PubChem Substance ID 57647647

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
grade   technical
assay   ≥97.0% (T)
mp   ≥300 °C (lit.)
SMILES string   OS(O)(=O)=O.Nc1nc(N)c(N)c(O)n1
InChI   1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4)
InChI key   RSKNEEODWFLVFF-UHFFFAOYSA-N

Description

Application

4-Hydroxy-2,5,6-triaminopyrimidine sulfate salt may be used in chemical synthesis.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?