• USA Home
  • 17381 - 2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
17381 Aldrich

2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate

≥97.0% (CE)

Synonym: 1,3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate, DFIH

Purchase

Properties

assay   ≥97.0% (CE)
mp   162-167 °C
storage temp.   2-8°C

Description

Other Notes

Very reactive reagent for peptide coupling in solution and SPPS

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Imidazolium Derived Reagents

N-Acylimidazoles were recognized in the early 1950s as reactive intermediates suitable for the acylation of amino compounds. The search for better coupling reagents than DCC led to the development of...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 7.
Keywords: Acylations, Chemfiles, Esterifications, Peptide synthesis, Search

Related Content

COMU – Safer and More Efficient Peptide Coupling Reagent

Peptide synthesis relies heavily on efficient and reliable coupling reagents. A low tendency for racemization is a key requirement. This is especially true for solid phase peptide synthesis—quantitat...
Keywords: Building blocks, Calorimetry, Condensations, Coupling reactions, Epimerizations, PEGylations, Peptide synthesis, Racemizations, Solid phase peptide synthesis, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?